KIMBLE® KONTES® Inlet Adapters, DWK Life Sciences
KT183000-2440
- ST Joint:24/40
- Altura:80 mm
- Referencia:KT183000-2440
- Se ajusta al diá. de la tubería:⅜"
- Nº de proveedor:183000-2440
Designed for use as a vent or for the introduction of vacuum or pressure.
- Bottom inner [ST] joint
- Side hose connection
Features an inner Standard Taper joint at the bottom and hose connection on the side.