Women’s Basic Lab Coats, Encompass
White, 12 (ladies)
17788-138
Small Business Enterprise
: 47401-W10
Basic princess-style, single-breasted women’s lab coats feature five-button closure, mock belt, two patch pockets, one breast pocket, and set-in sleeves for an attractive look.
Comfortable 80% polyester/20% cotton poplin blend. Fabric is treated with a stain release for easy care. Garment length is 100.3 cm (39¹/₂").
- Clothing Size:12 (ladies)
- Color:White
- Cat. No.:17788-138
- Sleeve or leg length:Long
- Garment application:Laboratory
- Gender:For ladies
- Closure type:Buttons
- Fabric:80% polyester/20% cotton poplin blend