ContactUsLinkComponent
SP Bel-Art Large Capacity Multipurpose Trays, Polypropylene, Bel-Art, Bel-Art Products, a part of SP
SP Bel-Art Large Capacity Multipurpose Trays, Polypropylene, Bel-Art, Bel-Art Products, a part of SP
Catalog # 74950-088
SP Bel-Art Large Capacity Multipurpose Trays, Polypropylene, Bel-Art, Bel-Art Products, a part of SP
Catalog # 74950-088
Supplier Number:  F16200-0011
Specifications
  • Descripción:
    Tray with Handles
  • Referencia:
    74950-088
  • La x An x Al:
    32.4 x 26.7 x 10.8 cm (123/4 x 101/2 x 41/4")
  • Nº de proveedor:
    162000011
Specifications
About this item
Rugged, large capacity 8.5L (9qt.) trays may be used to contain ice, a water bath, glassware, or instruments for autoclaving.
  • Rugged durability
  • Polypropylene construction
  • Available with or without handles
  • Autoclavable
Inside corners are smooth and rounded for easy cleaning. Top rim adds rigidity and provides handholds for transport. Identical trays can nest for efficient storage. Trays are available with or without handles. Steam autoclavable at 121°C (250°F).