Orders LinkContactUsLinkComponent
Modular Spill Containment Platforms with Grating, Eagle Manufacturing
Two-Drum Platform, 6½", 51½", 5000 lbs., 26¼", 30 gal., 32 lbs.
  56618-544
 :  1632
Modular Spill Containment Platforms with Grating, Eagle Manufacturing
  56618-544
Modular Spill Containment Platforms with Grating, Eagle Manufacturing
Modular Spill Containment Platforms with Grating, Eagle Manufacturing
Two-Drum Platform, 6½", 51½", 5000 lbs., 26¼", 30 gal., 32 lbs.
$8,381.81
Each (1UN)
Economical, low-profile platform units support 208.2L (55gal.) drums, provide incidental spill containment, and connect to form multiple configurations.
Constructed of chemical-resistant high-density polyethylene. Translucent tub is safety yellow with black grating supports. Grating has 4.4cm (13/4") square openings and removes easily for cleaning. Polyethylene ramp (56610-212) fits all platform units and simplifies drum placement.
 : One-drum platform includes one U-channel connector for joining units; Two- and four-drum platforms include two connectors.
  • Description:
    Two-Drum Platform
  • Height:
    6½"
  • Length:
    51½"
  • Load Capacity:
    5000 lbs.
  • Sump Capacity:
    30 gal.
  • Weight:
    32 lbs.
  • Width:
    26¼"