Connecting [ST] Adapters, Chemglass
Proveedor: CHEMGLASS INC.
Certificados
Acerca de este artículo
Connecting [ST] adapters are designed to connect dissimilar [ST] joint sizes.
- Reducing and enlarging adapters available
- Choose from a variety of [ST] joint sizes
For reducing adapters, lower inner joint is larger in size than the top outer joint. For enlarging adapters, lower inner joint is smaller in size than the top outer joint.
Información de pedido: For additional sizes, contact your Avantor sales representative.
Frequently Bought Together