KIMBLE® KONTES® Connecting [ST] Adapters for Distilling, DWK Life Sciences
Catálogo # KT167500-2440
Proveedor: DWK LIFE SCIENCES S. DE R.L. DE MX
Especificaciones
- ST Joint:24/40
- Anchura:105 mm
- Altura:155 mm
- Referencia:KT167500-2440
- Nº de proveedor:167500-2440
Especificaciones
Acerca de este artículo
Use connecting [ST] adapter as a component in atmospheric or vacuum distillations.
- Inner joints are at a 75° angle
- For use in distillation assemblies
Adapter KT167000-2440 has a [ST] top joint for a thermometer.