Orders LinkContactUsLinkComponent
Combitips® advanced Positive Displacement Pipette Tips, Eppendorf Quality
Combitips® advanced Positive Displacement Pipette Tips, Eppendorf Quality
   Opciones sostenibles disponibles
Supplier:   EPPENDORF BIO TOOLS
Combitips® advanced Positive Displacement Pipette Tips, Eppendorf Quality
   Opciones sostenibles disponibles
Certificates
Combitips® advanced Positive Displacement Pipette Tips, Eppendorf Quality
Combitips® advanced Positive Displacement Pipette Tips, Eppendorf Quality
9 Options Available Below
Product Details & Documents
Eppendorf Combitips® advanced, in 'Eppendorf Quality' grade, offer precise, positive displacement pipetting. Ideal for handling challenging liquids, ensuring accuracy and efficiency in diverse lab applications with their versatile and durable design.
  • Combitips® advanced tips are the dedicated consumable for all Repeater®/Multipette® multi-dispenser pipettes from Eppendorf
  • 9 volume sizes (0.1 to 50 ml) offer a maximum range of dispensing volumes, with increments as low as 100 nanoliters
  • Combitips® advanced in ‘Eppendorf Quality’ are reliable, easy-to-handle multi-dispenser tips for a variety of lab applications
  • Elongated Combitips® advanced in volumes 2.5, 5.0, and 10 ml will reach the bottom of most common laboratory vessels
  • Combitips® advanced are offered in five different purity grades to support a range of applications
  • An appropriate purity grade for each application: Eppendorf Quality, PCR clean, Forensic DNA Grade, sterile, or Biopur®
  • ACT® labeled by non-profit My Green Lab® to help make an informed choice regarding sustainability
The tip of choice when working with Repeater®/Multipette® multi-dispenser pipettes from Eppendorf:
Thanks to their revolutionary design, Combitips® advanced dispenser tips provide many benefits for work in a modern laboratory. With an integrated piston, they function according to the positive displacement principle and ensure that the correct volume of liquid is dispensed regardless of the physical properties of the liquid (e.g., viscosity, volatility, density, temperature). Achieve high-precision dispensing, independent of the flow characteristics of the liquid, and safely work with radioactive or toxic materials due to the hermetically sealed piston that prevents aerosol contamination.
Eppendorf pioneered the Repeater®/Combitips® multi-dispenser system in 1978 and improved it several times since. It was the very first pipetting system allowing users to fill the tip once and dispense multiple times. The concept remains a driver for efficient liquid handling and a staple in many labs working with plates or series of tubes. In 2023, the Combitips® advanced pipette tip line-up has been certified with ACT® Environmental Impact Factor Labels by the My Green Lab® non-profit organization, making it the ideal choice for labs seeking more sustainable products.
Certifications: ACT® Label and General quality certificates available, for lot-specific certificates please choose one of the other purity grades: PCR clean, Forensic DNA Grade, sterile, or Biopur®.
Caution: Please note: Combitips® advanced are designed for use with Eppendorf Repeater®/Multipette® multi-dispensers only.
Documents
Specifications for Product Family
Especificaciones
  • Catalog No:
  • 89232-930
  • 89232-932
  • 89232-934
  • 89232-936
  • 89232-938
  • 89232-940
  • 89232-942
  • 89232-944
  • 89232-946
  • Environmentally Preferable:
  • Low Manufacturing Impact|Product Transparency and Disclosure
  • Low Manufacturing Impact|Product Transparency and Disclosure
  • Low Manufacturing Impact|Product Transparency and Disclosure
  • Low Manufacturing Impact|Product Transparency and Disclosure
  • Low Manufacturing Impact|Product Transparency and Disclosure
  • Low Manufacturing Impact|Product Transparency and Disclosure
  • Low Manufacturing Impact|Product Transparency and Disclosure
  • Low Manufacturing Impact|Product Transparency and Disclosure
  • Low Manufacturing Impact|Product Transparency and Disclosure
  • Environmental certification:
  • Renewable Energy Certificate, ISO 14001, ACT Label
  • Renewable Energy Certificate, ISO 14001, ACT Label
  • Renewable Energy Certificate, ISO 14001, ACT Label
  • Renewable Energy Certificate, ISO 14001, ACT Label
  • Renewable Energy Certificate, ISO 14001, ACT Label
  • Renewable Energy Certificate, ISO 14001, ACT Label
  • Renewable Energy Certificate, ISO 14001, ACT Label
  • Renewable Energy Certificate, ISO 14001, ACT Label
  • Renewable Energy Certificate, ISO 14001, ACT Label
Especificaciones
Product Family Options
Información del producto
  • Volume
    Sterile
    Color Code
    Packed
    Availability
    Price
  • 0.1 ml
    -
    White
    100 pcs. (4 bags × 25 pcs.)
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (100UN)
    Case (100UN) - $4,290.00
    $4,290.00
    Case (100UN)
       $4,290.00
    Supplier Number: 0030089405
    Especificaciones:
    Product available on GSA Advantage®
      Low Manufacturing Impact|Product Transparency and Disclosure
    More
    Specifications
    Cat. No.
    89232-930
    Environmentally Preferable
    Low Manufacturing Impact|Product Transparency and Disclosure
    Environmental certification
    Renewable Energy Certificate, ISO 14001, ACT Label
    Product available on GSA Advantage®
      Low Manufacturing Impact|Product Transparency and Disclosure
  • 0.2 ml
    -
    Light Blue
    100 pcs. (4 bags × 25 pcs.)
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (100UN)
    Case (100UN) - $4,290.00
    $4,290.00
    Case (100UN)
       $4,290.00
    Supplier Number: 0030089413
    Especificaciones:
    Product available on GSA Advantage®
      Low Manufacturing Impact|Product Transparency and Disclosure
    More
    Specifications
    Cat. No.
    89232-932
    Environmentally Preferable
    Low Manufacturing Impact|Product Transparency and Disclosure
    Environmental certification
    Renewable Energy Certificate, ISO 14001, ACT Label
    Product available on GSA Advantage®
      Low Manufacturing Impact|Product Transparency and Disclosure
  • 0.5 ml
    -
    Violet
    100 pcs. (4 bags × 25 pcs.)
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (100UN)
    Case (100UN) - $4,290.00
    $4,290.00
    Case (100UN)
       $4,290.00
    Supplier Number: 0030089421
    Especificaciones:
    Product available on GSA Advantage®
      Low Manufacturing Impact|Product Transparency and Disclosure
    More
    Specifications
    Cat. No.
    89232-934
    Environmentally Preferable
    Low Manufacturing Impact|Product Transparency and Disclosure
    Environmental certification
    Renewable Energy Certificate, ISO 14001, ACT Label
    Product available on GSA Advantage®
      Low Manufacturing Impact|Product Transparency and Disclosure
  • 1.0 ml
    -
    Yellow
    100 pcs. (4 bags × 25 pcs.)
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (100UN)
    Case (100UN) - $4,290.00
    $4,290.00
    Case (100UN)
       $4,290.00
    Supplier Number: 0030089430
    Especificaciones:
    Product available on GSA Advantage®
      Low Manufacturing Impact|Product Transparency and Disclosure
    More
    Specifications
    Cat. No.
    89232-936
    Environmentally Preferable
    Low Manufacturing Impact|Product Transparency and Disclosure
    Environmental certification
    Renewable Energy Certificate, ISO 14001, ACT Label
    Product available on GSA Advantage®
      Low Manufacturing Impact|Product Transparency and Disclosure
  • 2.5 ml
    -
    Green
    100 pcs. (4 bags × 25 pcs.)
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (100UN)
    Case (100UN) - $4,290.00
    $4,290.00
    Case (100UN)
       $4,290.00
    Supplier Number: 0030089448
    Especificaciones:
    Product available on GSA Advantage®
      Low Manufacturing Impact|Product Transparency and Disclosure
    More
    Specifications
    Cat. No.
    89232-938
    Environmentally Preferable
    Low Manufacturing Impact|Product Transparency and Disclosure
    Environmental certification
    Renewable Energy Certificate, ISO 14001, ACT Label
    Product available on GSA Advantage®
      Low Manufacturing Impact|Product Transparency and Disclosure
  • 5.0 ml
    -
    Blue
    100 pcs. (4 bags × 25 pcs.)
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (100UN)
    Case (100UN) - $4,290.00
    $4,290.00
    Case (100UN)
       $4,290.00
    Supplier Number: 0030089456
    Especificaciones:
      Low Manufacturing Impact|Product Transparency and Disclosure
    More
    Specifications
    Cat. No.
    89232-940
    Environmentally Preferable
    Low Manufacturing Impact|Product Transparency and Disclosure
    Environmental certification
    Renewable Energy Certificate, ISO 14001, ACT Label
      Low Manufacturing Impact|Product Transparency and Disclosure
  • 10 ml
    -
    Orange
    100 pcs. (4 bags × 25 pcs.)
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (100UN)
    Case (100UN) - $4,290.00
    $4,290.00
    Case (100UN)
       $4,290.00
    Supplier Number: 0030089464
    Especificaciones:
    Product available on GSA Advantage®
      Low Manufacturing Impact|Product Transparency and Disclosure
    More
    Specifications
    Cat. No.
    89232-942
    Environmentally Preferable
    Low Manufacturing Impact|Product Transparency and Disclosure
    Environmental certification
    Renewable Energy Certificate, ISO 14001, ACT Label
    Product available on GSA Advantage®
      Low Manufacturing Impact|Product Transparency and Disclosure
  • 25 ml
    -
    Red
    100 pcs. (4 bags × 25 pcs.), each box containing one adapter
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (100UN)
    Case (100UN) - $5,420.00
    $5,420.00
    Case (100UN)
       $5,420.00
    Supplier Number: 0030089472
    Especificaciones:
    Product available on GSA Advantage®
      Low Manufacturing Impact|Product Transparency and Disclosure
    More
    Specifications
    Cat. No.
    89232-944
    Environmentally Preferable
    Low Manufacturing Impact|Product Transparency and Disclosure
    Environmental certification
    Renewable Energy Certificate, ISO 14001, ACT Label
    Product available on GSA Advantage®
      Low Manufacturing Impact|Product Transparency and Disclosure
  • 50 ml
    -
    Light grey
    100 pcs. (4 bags × 25 pcs.), each box containing one adapter
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (100UN)
    Case (100UN) - $5,420.00
    $5,420.00
    Case (100UN)
       $5,420.00
    Supplier Number: 0030089480
    Especificaciones:
    Product available on GSA Advantage®
      Low Manufacturing Impact|Product Transparency and Disclosure
    More
    Specifications
    Cat. No.
    89232-946
    Environmentally Preferable
    Low Manufacturing Impact|Product Transparency and Disclosure
    Environmental certification
    Renewable Energy Certificate, ISO 14001, ACT Label
    Product available on GSA Advantage®
      Low Manufacturing Impact|Product Transparency and Disclosure
Accessories
Información del producto
  • Catalog #
    Availability
    Price
  • Combitips advanced 50 mL adapter, Light Gray
    Combitips advanced 50 mL adapter, Light Gray
    89232-986
    Log in Para disponibilidad
    Each (1UN) - $805.00
Información del producto
  • Catalog #
    Availability
    Price
  • Combitips advanced 25 mL adapter, Red
    Combitips advanced 25 mL adapter, Red
    89232-984
    Log in Para disponibilidad
    Each (1UN) - $805.00