Orders LinkContactUsLinkComponent
epT.I.P.S.® Reloads, Pipette Tips
epT.I.P.S.® Reloads, Pipette Tips
   Opciones sostenibles disponibles
Supplier:   EPPENDORF BIO TOOLS
epT.I.P.S.® Reloads, Pipette Tips
   Opciones sostenibles disponibles
Certificates
epT.I.P.S.® Reloads, Pipette Tips
5 Options Available Below
Product Details & Documents
Eppendorf epT.I.P.S.® Reloads offer high-precision pipette tips in 'Eppendorf Quality' and 'PCR clean' purity grades. Eco-friendly, cost-effective, and designed for seamless compatibility with Eppendorf Box 2.0, they ensure accuracy and sustainability in labs.
  • Reduced waste when compared with disposable racks
  • Refill trays can be autoclaved up to 121 °C, in packaging and without foil, for subsequent use
  • Depending on tip size, the refill system is packaged as either dual-sided or in stack form
  • Available in purity grades of Eppendorf Quality and PCR clean
  • Color-coded trays for simple volume and pipette identification
  • ACT® labeled by non-profit My Green Lab® to help make an informed choice regarding sustainability
The perfect fit for Eppendorf pipettes: epT.I.P.S.® ensure a reliable pipetting system:
Make everyday work easier and more enjoyable with the epT.I.P.S.® (Eppendorf "Totally Integrated Pipetting System") pipette tips. These pipette tips are manufactured to extremely high quality and purity standards and are designed to cover all types of specific sampling needs. Offering reduced-force tip attachment and ejection with complete sealing, epT.I.P.S.® work in perfect combination with Eppendorf pipettes and form a fully integrated pipetting system that delivers maximum precision, accuracy and reproducibility. Eppendorf pipette tips with ideal wetting properties and optimum transparency are manufactured exclusively from high-purity plastic materials. The universal nose cone design allows Eppendorf tips to also be used with pipettes from other manufacturers.
How to recognize an original Eppendorf pipette tip?
All Eppendorf tips feature a 'ring' of drop-shaped relief elements on the edge of the tip crown.
Extended length for reliable sampling: epT.I.P.S.® L pipette tips:
The elongated pipette tips (epT.I.P.S.® L) enable reliable sample reach while pipetting from and into conical tubes, high reagent bottles, narrow deep vessels, cell culture flasks, or deepwell plates. The long and slim design of these elongated pipette tips allows free access to the sample with reduced risk of touching the sides of tubes or wells. Cross-contamination during pipetting can be reduced to a minimum. epT.I.P.S.® L are available in volumes of: 0.5 to 20 µl, 50 to 1250 µl, 0.2 to 5 ml, and 0.5 to 10 ml.
Certifications: Lot-specific certificates for the purity grade PCR clean can be downloaded from the Eppendorf website.
Specifications for Product Family
Especificaciones
  • Catalog No:
  • 47745-138
  • 47745-142
  • 47745-148
  • 47745-154
  • 47745-158
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
Especificaciones
Product Family Options
Eppendorf Quality (non-sterile)
Información del producto
  • Volume
    Length
    Color Code
    Packed
    Availability
    Price
  • 2 - 200 µl
    53 mm
    Yellow
    960 Tips (10 trays × 96 Tips)
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (960UN)
    Case (960UN) - $2,420.00
    $2,420.00
    Case (960UN)
       $2,420.00
    Supplier Number: 022491539
    Especificaciones:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    47745-138
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 50 - 1000 µl
    71 mm
    Blue
    960 Tips (10 trays × 96 Tips)
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (960UN)
    Case (960UN) - $2,710.00
    $2,710.00
    Case (960UN)
       $2,710.00
    Supplier Number: 022491555
    Especificaciones:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    47745-142
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
PCR Clean (non-sterile)
Información del producto
  • Volume
    Length
    Color Code
    Packed
    Availability
    Price
  • 0.1 - 10 µl
    34 mm
    Dark grey
    960 Tips (10 trays × 96 Tips)
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (960UN)
    Case (960UN) - $3,680.00
    $3,680.00
    Case (960UN)
       $3,680.00
    Supplier Number: 022491709
    Especificaciones:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    47745-148
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 2 - 200 µl
    53 mm
    Yellow
    960 Tips (10 trays × 96 Tips)
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (960UN)
    Case (960UN) - $2,930.00
    $2,930.00
    Case (960UN)
       $2,930.00
    Supplier Number: 022491733
    Especificaciones:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    47745-154
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 50 - 1000 µl
    71 mm
    Blue
    960 Tips (10 trays × 96 Tips)
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (960UN)
    Case (960UN) - $3,170.00
    $3,170.00
    Case (960UN)
       $3,170.00
    Supplier Number: 022491750
    Especificaciones:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    47745-158
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
    Product available on GSA Advantage®
      Low Manufacturing Impact