Orders LinkContactUsLinkComponent
epT.I.P.S.® Bulk - Pipette Tips
epT.I.P.S.® Bulk - Pipette Tips
   Opciones sostenibles disponibles
Supplier:   EPPENDORF BIO TOOLS
epT.I.P.S.® Bulk - Pipette Tips
   Opciones sostenibles disponibles
Certificates
epT.I.P.S.® Bulk - Pipette Tips
12 Options Available Below
Product Details & Documents
Eppendorf epT.I.P.S.® Bulk packs offer a practical, high-volume solution with 'Eppendorf Quality' pipette tips, ensuring precise and reliable measurements for various lab applications, packaged economically for high-throughput settings.
  • Original, high-quality Eppendorf pipette tips packaged in resealable bags
  • Available in sizes from 10 µl to 10 ml in Eppendorf Quality purity grade
  • epT.I.P.S.® Standard 200, 300 and 1000 µl tips are also available color-coded yellow and blue
  • epT.I.P.S.® Standard pipette tips can be autoclaved at 121 °C in 20 min
  • Fine retracted or extended tips simplify work with small volumes
  • ACT® labeled by non-profit My Green Lab® to help make an informed choice regarding sustainability
The perfect fit for Eppendorf pipettes: epT.I.P.S.® ensure a reliable pipetting system:
Make everyday work easier and more enjoyable with the epT.I.P.S.® (Eppendorf 'Totally integrated pipetting system') pipette tips. These pipette tips are manufactured to extremely high quality and purity standards and are designed to cover all types of specific sampling needs. Offering reduced-force tip attachment and ejection with complete sealing, epT.I.P.S.® work in perfect combination with Eppendorf pipettes and form a fully integrated pipetting system that delivers maximum precision, accuracy, and reproducibility. Eppendorf pipette tips with ideal wetting properties and optimum transparency are manufactured exclusively from high-purity plastic materials. The universal nose cone design allows Eppendorf tips to also be used with pipettes from other manufacturers.
How to recognize an original Eppendorf pipette tip?
All Eppendorf tips feature a 'ring' of drop-shaped relief elements on the edge of the tip crown.
Extended length for reliable sampling: epT.I.P.S.® L pipette tips:
The elongated pipette tips (epT.I.P.S.® L) enable reliable sample reach while pipetting from and into conical tubes, high reagent bottles, narrow deep vessels, cell culture flasks, or deepwell plates. The long and slim design of these elongated pipette tips allows free access to the sample with reduced risk of touching the sides of tubes or wells. Cross-contamination during pipetting can be reduced to a minimum. epT.I.P.S.® L are available in volumes of: 0.5 to 20 µl, 50 to 1250 µl, 0.2 to 5 ml, and 0.5 to 10 ml.
Certifications: General quality and ISO certificates available.
Documents
Specifications for Product Family
Especificaciones
  • Catalog No:
  • 47745-164
  • 47745-166
  • 47745-168
  • 47745-170
  • 47745-172
  • 47745-174
  • 47745-176
  • 89126-938
  • 47745-180
  • 47745-178
  • 47747-986
  • 47747-992
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
Especificaciones
Product Family Options
Información del producto
  • Volume
    Length
    Color Code
    Packed
    Availability
    Price
  • 0.1 - 10 µl
    34 mm
    Dark grey
    1000 Tips (2 bags×500 Tips)
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (1,000UN)
    Case (1,000UN) - $2,280.00
    $2,280.00
    Case (1,000UN)
       $2,280.00
    Supplier Number: 022492004
    Especificaciones:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    47745-164
    Environmentally Preferable
    Environmental certification
    Product available on GSA Advantage®
  • 0.1 - 20 µl
    40 mm
    Light orange
    1000 Tips (2 bags×500 Tips)
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (1,000UN)
    Case (1,000UN) - $2,280.00
    $2,280.00
    Case (1,000UN)
       $2,280.00
    Supplier Number: 022492012
    Especificaciones:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    47745-166
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 0.5 - 20 µl
    46 mm
    Light grey
    1000 Tips (2 bags×500 Tips)
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (1,000UN)
    Case (1,000UN) - $2,170.00
    $2,170.00
    Case (1,000UN)
       $2,170.00
    Supplier Number: 022492021
    Especificaciones:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    47745-168
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 2 - 200 µl
    53 mm
    Yellow
    1000 Tips (2 bags×500 Tips)
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (1,000UN)
    Case (1,000UN) - $1,410.00
    $1,410.00
    Case (1,000UN)
       $1,410.00
    Supplier Number: 022492039
    Especificaciones:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    47745-170
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 20 - 300 µl
    55 mm
    Orange
    1000 Tips (2 bags×500 Tips)
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (1,000UN)
    Case (1,000UN) - $1,410.00
    $1,410.00
    Case (1,000UN)
       $1,410.00
    Supplier Number: 022492047
    Especificaciones:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    47745-172
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 50 - 1000 µl
    71 mm
    Blue
    1000 Tips (2 bags×500 Tips)
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (1,000UN)
    Case (1,000UN) - $1,410.00
    $1,410.00
    Case (1,000UN)
       $1,410.00
    Supplier Number: 022492055
    Especificaciones:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    47745-174
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
  • 50 - 1250 µl
    76 mm
    Green
    1000 Tips (4 bags×250 Tips)
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (1,000UN)
    Case (1,000UN) - $2,390.00
    $2,390.00
    Case (1,000UN)
       $2,390.00
    Supplier Number: 022492063
    Especificaciones:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    47745-176
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 50 - 1250 µl
    103 mm
    Dark green
    1000 Tips (4 bags×250 Tips)
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (1,000UN)
    Case (1,000UN) - $2,420.00
    $2,420.00
    Case (1,000UN)
       $2,420.00
    Supplier Number: 022494018
    Especificaciones:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    89126-938
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 100 - 5000 µl
    120 mm
    Violet
    500 Tips (5 bags×100 Tips)
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (500UN)
    Case (500UN) - $1,920.00
    $1,920.00
    Case (500UN)
       $1,920.00
    Supplier Number: 022492080
    Especificaciones:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    47745-180
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
  • 250 - 2500 µl
    115 mm
    Red
    500 Tips (5 bags×100 Tips)
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (500UN)
    Case (500UN) - $1,650.00
    $1,650.00
    Case (500UN)
       $1,650.00
    Supplier Number: 022492071
    Especificaciones:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    47745-178
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 500 - 10000 µl
    165 mm
    Turquoise
    200 Tips (2 bags×100 Tips)
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (200UN)
    Case (200UN) - $1,970.00
    $1,970.00
    Case (200UN)
       $1,970.00
    Supplier Number: 022492098
    Especificaciones:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    47747-986
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 500 - 10000 µl
    243 mm
    Turquoise
    200 Tips (2 bags×100 Tips)
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (200UN)
    Case (200UN) - $2,810.00
    $2,810.00
    Case (200UN)
       $2,810.00
    Supplier Number: 022492101
    Especificaciones:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    47747-992
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
    Product available on GSA Advantage®
      Low Manufacturing Impact