Orders LinkContactUsLinkComponent
epT.I.P.S.® Racks - Pipette Tips
   Opciones sostenibles disponibles
Supplier:   EPPENDORF BIO TOOLS
epT.I.P.S.® Racks - Pipette Tips
   Opciones sostenibles disponibles
Certificates
epT.I.P.S.® Racks - Pipette Tips
4 Options Available Below
Product Details & Documents
Eppendorf epT.I.P.S.® Racks feature high-precision pipette tips in 'Eppendorf Quality', 'PCR clean', 'Biopur®', and 'Sterile' purity grades. All tips are supplied in disposible racks designed for optimal organization and user convenience in demanding laboratory environments.
  • Long, narrow tips reach the base of narrow and high vessels without touching the top of the vessel edge
  • Modern slim rack design with reclosable lid
  • Improved sustainability through 35% lower polypropylene consumption per rack
  • ACT® labeled by non-profit My Green Lab® to help make an informed choice regarding sustainability
The perfect fit for Eppendorf pipettes: epT.I.P.S.® ensure a reliable pipetting system:
Make everyday work easier and more enjoyable with the epT.I.P.S.® (Eppendorf 'Totally integrated pipetting system') pipette tips. These pipette tips are manufactured to extremely high quality and purity standards and are designed to cover all types of specific sampling needs. Offering reduced-force tip attachment and ejection with complete sealing, epT.I.P.S.® work in perfect combination with Eppendorf pipettes and form a fully integrated pipetting system that delivers maximum precision, accuracy, and reproducibility. Eppendorf pipette tips with ideal wetting properties and optimum transparency are manufactured exclusively from high-purity plastic materials. The universal nose cone design allows Eppendorf tips to also be used with pipettes from other manufacturers.
How to recognize an original Eppendorf pipette tip?
All Eppendorf tips feature a 'ring' of drop-shaped relief elements on the edge of the tip crown.
Extended length for reliable sampling: epT.I.P.S.® L pipette tips:
The elongated pipette tips (epT.I.P.S.® L) enable reliable sample reach while pipetting from and into conical tubes, high reagent bottles, narrow deep vessels, cell culture flasks, or deepwell plates. The long and slim design of these elongated pipette tips allows free access to the sample with reduced risk of touching the sides of tubes or wells. Cross-contamination during pipetting can be reduced to a minimum. epT.I.P.S.® L are available in volumes of: 0.5 to 20 µl, 50 to 1250 µl, 0.2 to 5 ml, and 0.5 to 10 ml.
 : Lot-specific certficates available for the purity grades PCR clean, Sterile, and Biopur® can be downloaded from the Eppendorf website.
Specifications for Product Family
Especificaciones
  • Catalog No:
  • 47745-198
  • 47745-072
  • 47745-074
  • 47745-076
  • 47745-078
  • 47747-990
  • 47751-669
  • 47751-661
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
Especificaciones
Product Family Options
Eppendorf quality (non-sterile)
Información del producto
  • Volume
    Length
    Color Code
    Packed
    Availability
    Price
  • 100 - 5000 µl
    120 mm
    Violet
    120 Tips (5 Racks×24 Tips)
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (120UN)
    Case (120UN) - $1,960.00
    $1,960.00
    Case (120UN)
       $1,960.00
    Supplier Number: 0030071638
    Especificaciones:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    47745-198
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
    Product available on GSA Advantage®
      Low Manufacturing Impact
Biopur® (sterile)
Información del producto
  • Volume
    Length
    Color Code
    Packed
    Availability
    Price
  • 0.1 - 20 µl
    40 mm
    Light orange
    480 Tips (5 Racks×96 Tips)
    Discontinued
    Supplier Number: 0030075226
    Especificaciones:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    47745-072
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 2 - 200 µl
    53 mm
    Yellow
    480 Tips (5 Racks×96 Tips)
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (480UN)
    Case (480UN) - $3,380.00
    $3,380.00
    Case (480UN)
       $3,380.00
    Supplier Number: 0030075234
    Especificaciones:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    47745-074
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 20 - 300 µl
    55 mm
    Orange
    480 Tips (5 Racks×96 Tips)
    Discontinued
    Supplier Number: 0030075242
    Especificaciones:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    47745-076
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 50 - 1000 µl
    71 mm
    Blue
    480 Tips (5 Racks×96 Tips)
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (480UN)
    Case (480UN) - $3,570.00
    $3,570.00
    Case (480UN)
       $3,570.00
    Supplier Number: 0030075250
    Especificaciones:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    47745-078
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 500 - 10000 µl
    165 mm
    Turquoise
    120 Tips (5 Racks×24 Tips)
    Discontinued
    Supplier Number: 0030075315
    Especificaciones:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    47747-990
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
    Product available on GSA Advantage®
      Low Manufacturing Impact
Sterile
Información del producto
  • Volume
    Length
    Color Code
    Packed
    Availability
    Price
  • 100 - 5000 µl
    120 mm
    Violet
    120 Tips (5 Racks×24 Tips)
    Discontinued
    Supplier Number: 0030071620
    Especificaciones:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    47751-669
    Environmentally Preferable
    Environmental certification
    Product available on GSA Advantage®
  • 20 - 300 µl
    55 mm
    Orange
    960 Tips (10 Racks×96 Tips)
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (960UN)
    Case (960UN) - $3,460.00
    $3,460.00
    Case (960UN)
       $3,460.00
    Supplier Number: 0030071573
    Especificaciones:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    47751-661
    Environmentally Preferable
    Environmental certification
    Product available on GSA Advantage®