ContactUsLinkComponent
Cleanroom NovaSponges, Micronova
Cleanroom NovaSponges, Micronova
Catálogo # 68400-071
Proveedor:  MICRONOVA MFG INC
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
undefined
Cleanroom NovaSponges, Micronova
Catálogo # 68400-071
Proveedor:  MICRONOVA MFG INC
Número de proveedor:  SP4-533
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Especificaciones
  • Descripción:
    NovaCel™ Sponge, white polyvinyl acetate
  • Grosor:
    7.6 cm (3")
  • Referencia:
    68400-071
  • An x La:
    7.6×12.7 cm (3×5")
  • Nº de proveedor:
    SP4-533
Especificaciones
Acerca de este artículo
Cleanroom sponges to meet every demand from isolators and laminar flow benches to equipment interiors and laboratory applications. Choose from a range of fabric covers for specialized cleaning.
  • Made with cleanroom approved materials
  • Durable, reusable and absorbent
  • Fabric covers securely sewn over reticulated urethane foam interiors
  • Shape and size options available
  • Available gamma irradiated for aseptic areas
  • Hold up to strong disinfectants
  • Autoclavable
NovaCel™ Sponge: Ultra-clean polyvinyl acetate, firm to the touch, but soft and pliant once wetted - ideal for lapping and polishing in wafer manufacture.
MegaTex® Sponge: Textured, thermally bonded, non-woven polyamide for textured surfaces, heavy soils, and viscous spills.
PolyMesh™ Sponges: 100% polyester with monofilament mesh - provides increased abrasion to remove photo resist, chemical deposits, and fluxes. Also used for fermentation tanks and veterinary laboratories.
NovaPoly™ Sponges: 100% smooth knit polyester - excellent disinfectant dispersal for a wide variety of cleanroom applications; FDA approved for indirect food contact.
NovaLite™ Sponges: latex bonded with rayon, chamois material, with 400% absorbency and great friction resistance - ideal for removing dust and fine powders in pharmaceutical, medical device and hi-tech cleanrooms. Excellent alternative to PVA where autoclaving is one of the criteria.
Microfiber Sponges: 80% polyester/ 20% polyamide material with superior cleaning ability and over 400% absorbency.
NovaSponges™: bonded, non-woven polyvinyl alcohol (PVA) - Soft and pliable, yet durable and hard wearing for wet or dry applications. Caution: Not recommended for use with isopropyl alcohol or chlorides.