SC-3 System Easy-On Hoods, WORKLON®
Supplier: SUPERIOR UNIFORM GROUP
Certificates
About this item
Reusable hoods are designed for use in microelectronics, semiconductor, disk drive, laser, and other industrial ISO Class 4 (FED-STD-209E Class 10/M2.5) cleanrooms
Hoods are made of static dissipative Burlington C3™ material. They drape across the shoulders and feature an adjustable snap front closure, rear horizontal snap adjustment, and two sets of vertical snaps for a facemask attachment. A color-coded hang loop denotes size.
Frequently Bought Together