WYPALL® X70 Wipers, KIMBERLY-CLARK PROFESSIONAL®
Supplier: KIMBERLY-CLARK CORP (WIPERS)
Certificates
About this item
WypAll® X70 Cloths are durable, reusable, absorbent, and stay strong for better cleaning performance.
- Fast absorbing HYDROKNIT® material is a hydroentangled cellulose/polypropylene blend with no adhesives or chemical binders
- Durable, rag-tough construction and textured surface
- Reusable, will not fall apart when wet
- Ideal for cleaning oil, dirt, grime, and solvents
- Picks up water almost instantly, and hangs on for fewer drips, outperforming laundered shop rags
Pound for pound absorbs more oil and water than rags.
Pound for pound absorbs more oil and water than rags.
Frequently Bought Together