Type I Safety Cans, Stainless Steel, Eagle Manufacturing
Supplier: EAGLE MFG CO
Certificates
About this item
For carrying and storing gasoline, oil, benzene, naphtha, and kerosene.
These safety cans reduce vapor explosion danger. Designed with a guard cap that protects pouring spout. Also features a free-swinging, carry-pour handle. Not for use with strong acids, hydroxides, or solvents.
Made of heavy 24-gauge stainless steel with a reinforced bottom. Equipped with Type 316 stainless steel flame arrester and fluoropolymer resin cap gasket. Bottom diameter: 28.5cm (111/4").
Certifications: Meet OSHA standards for safe industrial and construction practice. FM approved.
Frequently Bought Together