PYREX® 105° Vacuum Connecting Adapters with [ST] Joints, Corning
Catálogo # 89091-086
Proveedor: Corning
Product available on GSA Advantage®
Especificaciones
- ST Joint:19/22
- Environmentally Preferable:Low Manufacturing Impact
- Referencia:89091-086
- Nº de proveedor:8947-19
- Certificación ambiental:I-RECs, ISO 14001 - Monterrey
Especificaciones
Acerca de este artículo
This distillation adapter tube with sidearm has an outer joint at the top and an inner-sealed through drip tip joint at the bottom.
- 105° angle
- Top [ST] outer joint
- Bottom drip tip [ST] joint
- Serrated sidearm
Adapter tube features a [ST] outer joint at the top and an inner-sealed through drip tip [ST] joint at the bottom. The serrated sidearm is 10 mm OD.