Orders LinkContactUsLinkComponent
VWR® Cryogenic Protective Aprons
VWR® Cryogenic Protective Aprons
Supplier:   VWR International
VWR® Cryogenic Protective Aprons
Supplier:  VWR International
Certificates
VWR® Cryogenic Protective Aprons
3 Options Available Below
Product Details & Documents
These aprons offer ideal body and leg protection against splashes and spills during the filling and handling of liquid nitrogen supply cylinders and bulk tanks, and during maintenance of cryogenic equipment.
  • Provides durable protection when handling liquid nitrogen and servicing cryogenic equipment
  • Ideal when filling liquid nitrogen supply cylinders and bulk tanks
Multiple layers of 100% nylon and 3M™ insulation allow for durable protection in low-temperature environments. Adjustable straps with quick-release buckles provide added safety and convenience.
Documents
Specifications for Product Family
Especificaciones
  • Catalog No:
  • 97008-290
  • 97008-292
  • 97008-294
  • Garment application:
  • Medical|Industrial|Pharmaceutical
  • Medical|Industrial|Pharmaceutical
  • Medical|Industrial|Pharmaceutical
  • Risk area:
  • Cryo
  • Cryo
  • Cryo
  • Gender:
  • Unisex
  • Unisex
  • Unisex
  • Fabric:
  • 100% nylon and 3M™ insulation
  • 100% nylon and 3M™ insulation
  • 100% nylon and 3M™ insulation
Especificaciones
Product Family Options
Información del producto
  • L×W
    Availability
    Price
  • 91.4×61 cm (36×24")
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $3,417.19
    $3,417.19
    Each (1UN)
       $3,417.19
    Especificaciones:
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    97008-290
    Garment application
    Medical|Industrial|Pharmaceutical
    Risk area
    Cryo
    Gender
    Unisex
    Fabric
    100% nylon and 3M™ insulation
    Small Business Enterprise
  • 106.7×61 cm (42×24")
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $3,792.39
    $3,792.39
    Each (1UN)
       $3,792.39
    Especificaciones:
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    97008-292
    Garment application
    Medical|Industrial|Pharmaceutical
    Risk area
    Cryo
    Gender
    Unisex
    Fabric
    100% nylon and 3M™ insulation
    Small Business Enterprise
  • 122×61 cm (48×24")
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $4,537.67
    $4,537.67
    Each (1UN)
       $4,537.67
    Especificaciones:
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    97008-294
    Garment application
    Medical|Industrial|Pharmaceutical
    Risk area
    Cryo
    Gender
    Unisex
    Fabric
    100% nylon and 3M™ insulation
    Small Business Enterprise