ContactUsLinkComponent
Universal Inlet Adapters, Chemglass
Universal Inlet Adapters, Chemglass
Catálogo # 80065-980
Proveedor:  CHEMGLASS INC.
Small Business Enterprise
undefined
Universal Inlet Adapters, Chemglass
Catálogo # 80065-980
Proveedor:  CHEMGLASS INC.
Número de proveedor:  CG-1042-14
Small Business Enterprise
Especificaciones
  • ST Joint:
    14/20
  • Referencia:
    80065-980
  • Nº de proveedor:
    CG-1042-14
Especificaciones
Acerca de este artículo
These universal adapters have a thread at the top for a compression cap and o-ring.
  • Universal
  • Threaded top
  • Inner [ST] joint
Adapters feature a #7 em-Thread at the top for compression cap and Viton® O-ring.