WorkWipes® Series 60 Heavy-Duty Wipers, New Pig
89073-946
- Referencia:89073-946
- Empaquetado:Jumbo roll (1100 sheets)
- An x La:30.5x30.5 cm (12x12")
- Peso:6.7 kg (14.8 lbs.)
- Nº de proveedor:WIP661
Lightest of the heavy-duty WorkWipes® series, these economical wipers are made of cellulose and synthetic fibers for strength and low linting properties.
- Textured surface helps remove dirt and grime
- Ideal for applying disinfectants, degreasers, and solvents
Additional applications include grease removal, wiping hands, fast cleanups, and cleaning glass or stainless steel.