To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Referencia:15648-990
- Empaquetado:160 Wipes/Canister
- An x La:15.2 x 17 cm (6 x 63/4")
- Nº de proveedor:Q89072
Deep well lid provides ample space to store and access next wipe. Prominent contact time for easy identification and improved compliance with accrediation standards. Categorization of pathogens allows for quick identification of relevant efficacy claims. Safety icon clearly communicates key safety information on both label and lid.
- EPA-registered, low-level disinfectant
- Pre-moistened and ready-to-use
- Cleans and disinfects in one step
- Bactericidal and virucidal
- Compatible with a broad range of hard nonporous surfaces
- Reduces waste and saves time