To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Especificaciones
- Referencia:82020-458
- An x La:25.4 x 33 cm (10 x 13")
Especificaciones
Acerca de este artículo
These all-purpose wipes are versatile cloth-like wipes that are safe for skin contact, with no rinse required.
- Soft cloth-like material
- No rinse, skin formula
- Alcohol-free
A super absorbent layer of air-lay cellulose provides both softness and strength. Wipes are sensitive and soft enough for use on skin, and they are versatile enough for use on surfaces.