PROSAT® Presaturated Wipers in SatBox™ Containers, Contec®
33501-828
Small Business Enterprise
- Descripción:Polynit Heatseal Wipers, Refill for 33501-488
- Referencia:33501-828
- Empaquetado:100/Foil Pouch
- An x La:15.2 x 22.9 cm (6 x 9")
- Solvent:70% isopropyl alcohol, 30% deionized water
- Nº de proveedor:CSPS0001A
Premoistened knitted polyester wipers and nonwoven polypropylene wipers are saturated with various solvent blends.
- Made from high-density polypropylene
- Compatible with various blends of isopropyl alcohol, ethanol, and acetone
- Reduces solvent evaporation
Polynit Heatseal wipers are standard weight, textured 100% polyester interlock knit with nonabrasive, heatsealed edges. They are abrasion- and chemical-resistant. Economical nonwoven wipers are made of 100% meltblown polypropylene. PROSAT® wipers are ideal for wiping process tools and other equipment, as well as many other cleaning applications in critical environments. Color: white.
: Wipers are packaged in foil pouches except for wipers 33501-490, which are packaged in resealable pouches for easy dispensing and to preserve cleanliness and solvent saturation levels. Each case includes one SatBox™ container, except for refill wipers 33501-828. Pouches are individually lot-numbered for traceability.