PROSAT® Sterile Presaturated Nonwoven Wipers, Contec®
Meltblown polypropylene wipes provide a consistent release of solvent to thoroughly remove surface contaminants in critical environments. PROSAT® presaturated wipes are exceptionally clean and also free from additives of any kind. This particular meltblown polypropylene contains very low levels of sodium and other ions.
- Suitable for Grade A/B or ISO Class 5 environments
- Validated sterile per gamma irradiation
- Low in particles and fiber; ideal for product contact areas
- Microsized filaments provide exceptional particle removal
- Especially suitable for the metered release of 70% alcohol
- Reduces VOC emissions and solvent use
- Resealable pouch preserves cleanliness and solvent saturation levels
PROSAT® meltblown polypropylene wipes are ideal for wiping articles prior to pass through, cleaning process tools and other equipment, and general surface cleaning. Meltblown polypropylene fabric is exceptionally clean and free from additives.
Presaturated wipes ensure consistent saturation of each wipe independent of operator. The wipes are provided in convenient and easy to use peel and reseal pouches.
- Catalog No:
- 10014-422
- 10024-280
- 68310-404
- Material:
- PP
- PP
- PP
- Peso base:
- 36 g/m²
- 36 g/m²
- 34 g/m²
- Extraíbles:
- 0.01 g/m² using deionized water, 0.07 g/m² using isopropyl alcohol
- 0.01 g/m² using deionized water, 0.07 g/m² using isopropyl alcohol
- 0.040 g/m² using deionized water, 0.020 g/m² using isopropyl alcohol
- Fibras (>100 µm):
- 6.3×10³/m²
- 6.3×10³/m²
- 0.145×10³/m²
- Particle generation (>=0,5 µm):
- 5.8×10⁶/m²
- 5.8×10⁶/m²
- 23.9×10⁶/m²
- Iones, cloruro:
- 5.0 ppm
- 5.0 ppm
- 8.6 ppm
- Iones - sodio:
- 3.0 ppm
- 3.0 ppm
- 0.08 ppm
Frequently Bought Together