Phlebotomy Containers, Covidien
946 ml (32 oz.), 15 x 10 x 11 cm (6 x 4 x 41/4"), Red
Supplier #: 8900SA
These containers are ideal for the disposal of blood, hypodermic needles, and I.V. needles. Units are portable and are compatible with present blood drawing trays and techniques. Supplied with clear lids.
Containers meet or exceed JCAHO, OSHA, and NIOSH/CDC standards and requirements.
needles. Units are portable and are compatible with present blood drawing trays and techniques. Supplied with clear lids.
Ordering information: Rainbow collection (15704-230) includes 33 red, 33 yellow, and 34 purple containers.
Documents
- Capacity:946 ml (32 oz.)
- Color:Red
- L×D×H:15 x 10 x 11 cm (6 x 4 x 41/4")