SCIENCEWARE®, Biohazard Sharp Object Pouch and Stand, Bel-Art
Acerca de este artículo
This solid paperboard pouch is 10 mil thick and offers protection against sharp-edged objects prior to and during disposal. It remains open in an oval shape by folding the bottom flaps. After filling, the top flaps close and lock in place for safe disposal. Printed with biohazard symbol. Poxygrid® Safety Pouch Stand (11215-847), made of welded steel wire with long-lasting epoxy coating, keeps pouch standing upright for easy access.
- Protects against sharp-edged objects for disposal
- Top flaps close and lock in place
- Epoxy-coated welded steel wire stand provides easy, upright access
It remains open in an oval shape by folding the bottom flaps. After filling, the top flaps close and lock in place for safe disposal. Printed with biohazard symbol. Poxygrid® Safety Pouch Stand (11215-847), made of welded steel wire with long-lasting epoxy coating, keeps pouch standing upright for easy access.
2 Options Available Below
Product Details & Documents
This solid paperboard pouch is 10 mil thick and offers protection against sharp-edged objects prior to and during disposal. It remains open in an oval shape by folding the bottom flaps. After filling, the top flaps close and lock in place for safe disposal. Printed with biohazard symbol. Poxygrid® Safety Pouch Stand (11215-847), made of welded steel wire with long-lasting epoxy coating, keeps pouch standing upright for easy access.
- Protects against sharp-edged objects for disposal
- Top flaps close and lock in place
- Epoxy-coated welded steel wire stand provides easy, upright access
It remains open in an oval shape by folding the bottom flaps. After filling, the top flaps close and lock in place for safe disposal. Printed with biohazard symbol. Poxygrid® Safety Pouch Stand (11215-847), made of welded steel wire with long-lasting epoxy coating, keeps pouch standing upright for easy access.
Documents
11215-836
Imágenes 360
Bel-Art Scienceware® Tools for Biohazard Disposal
Documentación del producto
Product Family Options
Información del producto