SatPax® 1000, Presaturated Wipers, Berkshire
Proveedor: BERKSHIRE CORP
Certificados
Acerca de este artículo
SatPax® wipers provide a convenient combination of solvent and wiper that improves the cleaning process.It resolves safety issues associated with storage, mixing, and dispensing of solvents.
- Low extractables and fiber and particle counts
- Smooth and durable with good wet strength
- Reduces alcohol usage and preparation/handling costs
- Increases cleaning efficiency
SatPax® 1000 nonwoven polyester/cellulose wipers are saturated with semiconductor grade isopropyl alcohol and deionized water. Knitted polyester wipers are cleanroom laundered and are presaturated with a blend of semiconductor grade isopropyl alcohol and reverse osmosis deionized water filtered to 0.1 µm.
Certificaciones: ISO Class 5 – 8. Cleanroom Class 100 – 100 000. EU Grade B/C/D. USP 797.
Información de pedido: Wipers are folded and packaged in a resealable cleanroom compatible pouch (containing C-folded wipers) and canisters (containing a roll of perforated wipers).
Información relacionada
AT-SatPax 1000_Canister AT-SatPax1000_Pouch
Hoja de datos
VWR GHS Compliance Letter (US)
Detalles adicionales del producto
Frequently Bought Together