Indoor Plastic Wastebaskets, Rubbermaid®
89073-322
- Profundidad:11"
- Capacidad:39 L (10.3 gal.)
- Color:Gray
- Altura:19⅞
- Matriz:Plastic
- Referencia:89073-322
- Forma:Rectangular Prism
- Para ser utilizado con:Rubbermaid® Rectangular Free-Swinging Lids
- Nº de proveedor:RCP 2957 GRA
- Lid Type:Rubbermaid® Commercial 306700
Wastebaskets fit under standard desk height. Low-density polyethylene construction prevents chipping, rusting, and denting. Rolled rims increase strength and make handling and cleaning easier.
- For office waste disposal
- All-plastic construction won't chip, rust, or dent
- Fits under standard desk heights, even with extended lid
- Rolled rims add strength
- Opens on top
- easy-to-clean
This waste basket fits under a standard desk height, even when the swing top is fully extended.