Bluesorb® 750 Wipers, Berkshire
Proveedor: BERKSHIRE CORP
Certificados
Acerca de este artículo
All-purpose Class 100 wipers are extremely absorbent.
- Excellent combination of the synthetic polyester strength and cleanliness with the absorbent characteristics of cellulose
- Blue color allows differentiation for guarding against migration into critical areas
- Smooth, absorbent, and durable
- Autoclavable
- Not recommended for applications where 100% water is used; material may not be colorfast
Made of non-woven polyester/cellulose. Excellent dry and wet strength. Blue color provides an instant absorption indicator.
Especificaciones
- Catalog No:
- 21914-707
- 21914-740
- Peso base:
- 67.1 g/m²
- 67.1 g/m²
- Absorbancia, extrínseca:
- 309 cc/m²
- 309 cc/m²
- Absorbancia, intrínseca:
- 4.6 cc/g
- 4.6 cc/g
- Absorbancia, velocidad:
- 3 sec.
- 3 sec.
- Extraíbles:
- 0.18% using deionized water, 0.040% using isopropyl alcohol
- 0.18% using deionized water, 0.040% using isopropyl alcohol
- Fibras (>100 µm):
- 110/cm²
- 110/cm²
- Particle generation (>=0,5 µm):
- Dry flex test: 1300/ft.³
- Dry flex test: 1300/ft.³
- Particle generation (>=0,5 µm) - biaxial shake:
- 22,000/cm²
- 22,000/cm²
- Iones - calcio:
- 260 ppm
- 260 ppm
- Iones - magnesio:
- 29 ppm
- 29 ppm
- Iones - potasio:
- 3.0 ppm
- 3.0 ppm
- Iones - sodio:
- 14 ppm
- 14 ppm