PYREX® Milk Dilution Bottles, with Screw Cap, Corning
Proveedor: Corning
Certificados
Acerca de este artículo
Made of PYREX® borosilicate glass. Used for general storage of solutions and tissue culture work.
- Wide mouth bottle offers easy measuring or filling
- GPI 28-400 threaded black phenolic screw cap, supplied unattached, has a cemented-in rubber liner and is suitable for autoclaving
- With a cut-line graduation mark at 99 ±1 ml
These bottles are designed from the requirements for milk dilution bottles given in the 'Standard Methods for the Examination of Dairy Products' published by the American Public Health Association.
Especificaciones
- Catalog No:
- 16163-026
- 16163-004
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Certificación ambiental:
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey