Amplitude™ Epsilon™ Wipes, Contec®
Amplitude™ Epsilon™ Wipes, Flat Stacked, 300/Bag, 12 Bags/Case, 22.9×22.9 cm (9×9")
Supplier: CONTEC, INC. MX
: AMEP0001
Jump to:
Product Details & Documents
Amplitude™ Epsilon™ Wipes are made of a special grade blue cellulose and polyester hydroentangled to form a wipe with improved cleanliness and excellent sorbency.
- Recommended for ISO Class 5 to 6 environments
- Dye-fast blue color allows for differentiation between wipes used in different areas
- Double packaged to preserve product integrity
The cellulose uses a dye fast color, to minimize color bleeding. Blue color allows for differentiation in critical areas. Ideal for use in wet areas requiring spill pickup and cleanup.
Especificaciones
- Description:Amplitude™ Epsilon™ Wipes, Flat Stacked
- Packaging:300/Bag, 12 Bags/Case
- W×L:22.9×22.9 cm (9×9")