Amplitude™ Sigma™ Wipes
Supplier: CONTEC, INC. MX
Jump to:
Acerca de este artículo
Amplitude™ Sigma™ Wipes are a hydroentangled blend of 55% cellulose and 45% polyester. They are highly sorbent, offering excellent durability and good cleanliness.
- Recommended for ISO Class 5 to 8 environments
- Hydroentangled fabric provides low levels of particles and extractables
- Excellent for wiping and absorbing spills
- Strong and durable both wet and dry
- Cost-effective general-purpose wipe
Amplitude™ Sigma™ wipes are strong and durable wet or dry, making them excellent for use in general cleaning and spill pickup.
2 Options Available Below
Product Details & Documents
Amplitude™ Sigma™ Wipes are a hydroentangled blend of 55% cellulose and 45% polyester. They are highly sorbent, offering excellent durability and good cleanliness.
- Recommended for ISO Class 5 to 8 environments
- Hydroentangled fabric provides low levels of particles and extractables
- Excellent for wiping and absorbing spills
- Strong and durable both wet and dry
- Cost-effective general-purpose wipe
Amplitude™ Sigma™ wipes are strong and durable wet or dry, making them excellent for use in general cleaning and spill pickup.
Documents
Product Family Options
Información del producto
- PackagingW×LAvailabilityPrice
- 89030-186Amplitude™ Sigma™ Wipes, flat stackedAmplitude™ Sigma™ Wipes, flat stacked300/bag, 12 bags/case22.9×22.9 cm (9×9")Paquete de 300$0.00Case de 12 (300UN)$7,522.68Supplier Number: AMSI0001
- 89030-188Amplitude™ Sigma™ Wipes, flat stackedAmplitude™ Sigma™ Wipes, flat stacked150/bag, 18 bags/case30.5×30.5 cm (12×12")Paquete de 150$0.00Case de 18 (150UN)$10,892.88Supplier Number: AMSI0002