KIMTECH PURE® W4 Critical Task Wipers, Kimberly-Clark Professional®
Catalog # 21908-205
Supplier: KIMBERLY-CLARK CORP (WIPERS)
Specifications
- Referencia:21908-205
- Empaquetado:100/Bag
- An x La:30.5 x 30.5 cm (12 x 12")
- Nº de proveedor:33330
Specifications
About this item
Low-linting wipers for ISO Class 4 (FED-STD-209E Class 10/M2.5) cleanroom assembly operations, pharmaceutical operations, and other clean environments.
- 100% polypropylene, non-woven fabric is low in particulate matter and extractable levels
- Soft, absorbent, durable
- Contain no binders or reinforcing fibers
- Eliminiate surface fibers, trap particles, will not scratch surfaces
- Color: white
Each wiper can absorb over four times its weight in liquids. Acid-, base-, and solvent-resistant; will not break down or disintegrate when exposed to chemicals.
For ISO Class 4 (FED-STD-209E Class 10/M2.5) cleanroom assembly operations, pharmaceutical operations, and other clean environments.
Certifications: Conform to EIA 541 standard. Meet the USDA requirements for food contact.
Ordering information: Packaged in antistatic, self-dispensing double poly bags.