Corning® Disposable Anti-static Spatulas, Corning
Supplier: Corning
Certificates
About this item
These spatulas are designed to save time and to provide contamination-free samples. They eliminate the recycling and resterilizing necessary with reusable spatulas.
- Antistatic
- Sterile
- RNase-/DNase-free
- Nonpyrogenic
Each spatula features two different tips to meet a variety of sampling requirements.
Ordering information: Available in five different configurations. Microspatulas available in two configurations.
Packaging: Individually packaged.
Specifications
- Catalog No:
- 38888-610
- 77776-848
- 77776-850
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Certificación ambiental:
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa