Orders LinkContactUsLinkComponent
Sample Bottles, Wide Mouth, Wheaton
Sample Bottles, Wide Mouth, Wheaton
Sample Bottles, Wide Mouth, Wheaton
Sample Bottles, Wide Mouth, Wheaton
Sample bottles are designed for collecting, storing, and exhibiting a wide variety of specimens in the lab or field. PE caps snap over a special sealing ring to form an effective barrier to moisture, making these bottles especially suited for hygroscopic materials. Manufactured from Wheaton 800 soda-lime flint glass. Packed in convenient Lab File® cases.
  • Manufactured from Wheaton 800 soda-lime flint glass
PE caps snap over a special sealing ring to form an effective barrier to moisture, making these bottles especially suited for hygroscopic materials.
 : Packed in convenient Lab File® cases.
Product Family Options
  • Capacity
    Cap Size
  • 16 ml (0.54 oz.)
    30 mm
    Case (144UN)
    Case (144UN) - $5,250.00
    $5,250.00
    Case (144UN)
       $5,250.00
     : 225536
  • 24 ml (0.8 oz.)
    30 mm
     : 225538
  • 30 ml (1 oz.)
    34 mm
    Case (72UN)
    Case (72UN) - $8,260.35
    $8,260.35
    Case (72UN)
       $8,260.35
     : 225543
  • 60 ml (2 oz.)
    45 mm
     : -