AC Round Bottles, WHEATON®, DWK Life Sciences
500 ml (16.9 oz.), 70-400, 76×145 mm, Convenience Packed, Glass, White PP cap with polyvinyl liner
Supplier: DWK LIFE SCIENCES S. DE R.L. DE MX
Supplier #: W216999
AC Round Bottles, WHEATON®, DWK Life Sciences
Catalog # 14236-010
Supplier: DWK LIFE SCIENCES S. DE R.L. DE MX
Jump to:
Product Details & Documents
Atlantic City round bottles are ideal for sampling use and can also be used to store dry powders, soil samples, and specimens.
- Manufactured from Wheaton '800' glass
Certifications: Meets USP Type III standards.
Specifications
- Cap Size:70-400
- Cap Type:White PP cap with polyvinyl liner
- Capacity:500 ml (16.9 oz.)
- Material:Glass
- Packaging:Convenience Packed
- Diameter ×H:76×145 mm