Orders LinkContactUsLinkComponent
MicroFirst® Wipers, Berkshire
MicroFirst® Wipers, Berkshire
22.9 x 22.9 cm (9 x 9")
Catalog # 21913-192
Small Business Enterprise
Supplier:   BERKSHIRE CORP
Número de proveedor #:  MF.0909.20
MicroFirst® Wipers, Berkshire
Catalog # 21913-192
Supplier:  BERKSHIRE CORP
Small Business Enterprise
MicroFirst® Wipers, Berkshire
MicroFirst® Wipers, Berkshire
22.9 x 22.9 cm (9 x 9")
Price
$1,110.00
Log in Para disponibilidad
Paquete de 300 Case de 20 (300UN)
Product Details & Documents
Nonwoven wipers engineered for applications in ISO Class 5 (FED-STD-209E Class 100/M3.5) environments, including spill control and surface cleaning. Made of nonwoven, spunlaced DuPont™ Sontara® Micropure 100 fabric, a polyester/cellulose blend made without binders or surfactants. Apertured construction allows for increased absorbency. Excellent particulate and purity control.
  • Excellent combination of the synthetic polyester strength and cleanliness with the absorbent characteristics of cellulose
  • Apertured for increased absorbency and increased surface texture for improved cleaning efficiency
  • Low extractable levels and particle and fiber counts
  • Very durable with high wet strength
Made of nonwoven, spunlaced DuPont™ Sontara® Micropure 100 fabric, a polyester/cellulose blend made without binders or surfactants. Apertured construction allows for increased absorbency. Excellent particulate and purity control.
Ordering information: Wipers are double-wrapped for optimal cleanliness.
Documents
Especificaciones
  • W×L:
    22.9 x 22.9 cm (9 x 9")
  • Extractables:
    0.68% using deionized water, 0.031% using isopropyl alcohol
  • Cat. No.:
    21913-192
  • Absorbency - extrinsic:
    278 ml/m²
  • Absorbency - intrinsic:
    4.9 ml/g
  • Absorbency rate:
    3 sec.
  • Fibres (>100 µm):
    68/cm²
  • Basis weight:
    57 g/m²
  • Ions - sodium:
    14 ppm
  • Particle generation (>=0,5 µm) - biaxial shake:
    18 x 10³/cm²
  • Ions - magnesium:
    6.3 ppm
  • Ions - potassium:
    4.3 ppm
  • Ions - calcium:
    24 ppm