Acerca de este artículo
These nonwoven wipers are suitable for use in critical environments, for absorbing spills of water and most common solvents, and for tasks such as cleaning laminar flow benches. Hydroentangled blends produce durable wipers that pick up liquids quickly. Low extractables and metallic ions make these wipers good for use where chemical purity is of prime importance. Spec-Wipe® 3 wipers are made from a blend of 45% polyester and 55% cellulose fibers. Spec-Wipe® 3e wipers are made from a creped fabric that is a blend of 46% polyester and 54% cellulose fibers. The creped fabric provides extra softness and bulk.
- Made from a blend of 45% polyester and 55% cellulose fibers
- For use in critical environments
- Low extractables
- Creped fabric
Hydroentangled blends produce durable wipers that pick up liquids quickly.
Low extractables and metallic ions make these wipers good for use where chemical purity is of prime importance.
Especificaciones
- Catalog No:
- 21912-042
- 21914-758
- Modelo:
- Spec-Wipe® 3
- Spec-Wipe® 3
- Peso base:
- 67 g/m²
- 67 g/m²
- Absorbancia:
- 335 ml/m²
- 335 ml/m²
- Absorbancia, velocidad:
- 1 sec.
- 1 sec.
- Extraíbles:
- 0.056 g/m² using deionized water, 0.015 g/m² using isopropyl alcohol
- 0.056 g/m² using deionized water, 0.015 g/m² using isopropyl alcohol
- Fibras (>100 µm):
- 14.8×103/m²
- 14.8×103/m²
- Particle generation (>=0,5 µm):
- 17.1 x 10⁶/m²
- 17.1 x 10⁶/m²
- Iones, cloruro:
- 7.8 ppm
- 7.8 ppm
- Iones - sodio:
- 11.2 ppm
- 11.2 ppm
Especificaciones
Información relacionada
Certificados
Buscar por número de loteAvantor Top Rated Guide Production Insight Solutions for Controlled Environments and Manufacturing Processes VWR Production insight V1 VWR Production Supplies Brochure VWR Spec-Wipe 3 Wipers
Documentación del producto
VWR Spec-Wipe 3 Wipers VWR Spec-Wipe 3e Wipers