Vectra® Alpha® 10 and Alpha® 10 Cleanroom Wipers, Texwipe®
TWTX1010
: Texwipe
- Descripción:Vectra® Alpha® 10
- Fibras (>100 µm):400/m²
- Extraíbles:0.02 g/m² using deionized water, 0.06 g/m² using isopropyl alcohol
- Peso base:135 g/m²
- Particle generation (0,5 - 5 µm) - ASTM E2090 test:9.6 x 10⁶/m²
- Particle generation (5 - 100 µm) - ASTM E2090 Test:2 x 10⁵/m²
- Absorbancia, extrínseca:300 ml/m²
- Absorbancia, velocidad:0.5 sec.
- Absorbancia, intrínseca:2.22 ml/g
- Iones - potasio:0.08 ppm
- Iones - sodio:0.25 ppm
- Iones, cloruro:0.45 ppm
- Referencia:TWTX1010
- An x La:22.9 x 22.9 cm (9 x 9")
- Nº de proveedor:TX1010
These 100% continuous-filament polyester wipers are constructed in a double-knit, no-run, interlock pattern that is excellent for cleaning process equipment.
- All four cut edges of the wiper are thermally sealed
- Border seal provides a protective edge around the body of each wiper for minimum particle generation
Suitable for use in ISO Class 3 (FED-STD-209E Class 1/M1.5) environments. To reduce particulation, wiper TWTX1010 is Vectra processed and wiper TWTX1012 is laundered in an ISO Class 4 (FED-STD-209E Class 10/M2.5) cleanroom.
: Wipers are packed in SolventSafe Bag-Within-A-Bag® packaging.