Vectra® AlphaSorb® 10 and AlphaSorb® 10 Wipers, ITW Texwipe®
Catalog # TW-TX1052
Supplier: Texwipe
Specifications
- Descripción:AlphaSorb® 10
- Fibras (>100 µm):1000/m²
- Extraíbles:0.02 g/m² using deionized water, 0.10 g/m² using isopropyl alcohol
- Peso base:195 g/m²
- Particle generation (0,5 - 5 µm) - ASTM E2090 test:13 x 10⁶/m²
- Particle generation (5 - 100 µm) - ASTM E2090 Test:2.25 x 10⁵/m²
- Absorbancia, extrínseca:540 ml/m²
- Absorbancia, velocidad:0.5 sec.
- Absorbancia, intrínseca:2.8 ml/g
- Iones - potasio:0.13 ppm
- Iones - sodio:0.35 ppm
- Iones, cloruro:0.09 ppm
- Referencia:TW-TX1052
- An x La:30.5 x 30.5 cm (12 x 12")
- Nº de proveedor:TX1052
Specifications
About this item
Two-ply double-knit 100% continuous-filament polyester wipers offer high sorbency performance. All four edges are thermally sealed, forming a border to prevent particle and fiber release. Suitable for use in ISO Class 3 (FED-STD-209E Class 1/M1.5) environments. To reduce particulation, wiper TWTX1050 is Vectra processed and wiper TW-TX1052 is laundered in an ISO Class 4 (FED-STD-209E Class 10/M2.5) cleanroom.
- All four edges are thermally sealed, forming a border to prevent particle and fiber release
Suitable for use in ISO Class 3 (FED-STD-209E Class 1/M1.5) environments. To reduce particulation, wiper TWTX1050 is Vectra processed and wiper TW-TX1052 is laundered in an ISO Class 4 (FED-STD-209E Class 10/M2.5) cleanroom.
Ordering information: SolventSafe Bag-Within-A-Bag® packaging.