Polynit Heatseal Wipes, Contec®
Proveedor: CONTEC, INC. MX
Certificados
Acerca de este artículo
For general-purpose cleaning in ISO Class 3 (FED-STD-209E Class 1/M1.5) environments. Ideal for wiping exterior surfaces of products and cleaning interior surfaces of process tools or other equipment.
- Recommended for ISO Class 3–5 environments for nonsterile wipes
- Recommended for Grade A/B or ISO Class 5 environments for sterile wipes
- Laundered knitted 100% polyester fabric for extremely low levels of particles and fibers
- Heat sealed edges help prevent particle contamination
- 100% knitted polyester is abrasion and chemical resistant
- Laser-cut, sealed edge prevents release of fibers
Available square or as HexaWipe™ wipers with six heat-sealed edges. Hexagonal shape readily distinguishes critical-use wipers from other, noncritical-use wipers.
The sterile wipes are gamma irradiated and validated per AAMI guidelines to a 10⁻⁶ SAL. They can be used in the highest grade life science cleanrooms.
Información relacionada
Frequently Bought Together