ContactUsLinkComponent
Polynit Wipers, Contec®
Polynit Wipers, Contec®
Catalog # 68310-166
Supplier:  CONTEC, INC. MX
Small Business Enterprise
undefined
Polynit Wipers, Contec®
Catalog # 68310-166
Supplier:  CONTEC, INC. MX
Supplier Number:  PN-44
Small Business Enterprise
Specifications
  • Descripción:
    Polynit Wipes, Flat Stacked
  • Fibras (>100 µm):
    2.4×10³/m²
  • Extraíbles:
    0.0045 g/m² using deionized water, 0.0224 g/m² using isopropyl alcohol
  • Peso base:
    152 g/m²
  • Particle generation (>=0,5 µm):
    4.0 x 10⁶/m²
  • Absorbancia, extrínseca:
    417.3 ml/m²
  • Absorbancia, velocidad:
    <1 sec.
  • Absorbancia, intrínseca:
    2.89 ml/g
  • Iones - sodio:
    None detected (<0.20 ppm)
  • Iones, cloruro:
    None detected (<0.20 ppm)
  • Referencia:
    68310-166
  • Empaquetado:
    300/Bag, 16 Bags/Case
  • An x La:
    10.2×10.2 cm (4×4")
  • Nº de proveedor:
    PN-44
Specifications
About this item
For general-purpose cleaning in ISO Class 5 (FED-STD-209E Class 100/M3.5) environments. Ideal for wiping exterior surfaces of products and cleaning interior surfaces of process tools or other equipment.
  • Recommended for ISO Class 5 to 6 environments
  • Laundered knitted 100% polyester fabric
  • Soft edge is ideal for use on surfaces which are vulnerable to scratching
  • 100% knitted polyester is compatible with many solvents
  • Durable fabric is abrasion and chemical resistant
Polyester wipes are chemically resistant and exceptionally low in particles and fibers. An interlock knit creates a durable fabric, the addition of a periodic additional stitch of a no-run interlock knit prevents the fabric unraveling creating an even more durable fabric with strong edges.
The wipes are laundered and packaged in an ISO Class 4 cleanroom. They have good sorbency with solvents and are abrasion and chemical resistant. A validated sterile version is available.