Orders LinkContactUsLinkComponent
Diamond Scriber
Diamond Scriber
Diamond scriber, 152 mm (6")
Catalog # 52865-005
Número de proveedor #:  200-22A-100
Diamond Scriber
Catalog # 52865-005
Diamond Scriber
Diamond Scriber
Diamond scriber, 152 mm (6")
Price
$566.28
Log in Para disponibilidad
Each (1UN)
Product Details & Documents
For permanently marking glass slides, test tubes, and laboratory glassware
Sharp, selected diamond mounted in a round, clear aluminum handle tapered to fit hand comfortably. Tip retracts for protection and safety.
Ordering information: Also available in blue, black, red, and silver handles; contact your Avantor sales representative for more information.
Especificaciones
  • Description:
    Diamond scriber
  • Length:
    152 mm (6")
Accessories
Información del producto
  • Catalog #
    Availability
    Price
  • Replacement point
    Replacement point
    52865-118
    Log in Para disponibilidad
    Each (1UN) - $235.37