ContactUsLinkComponent
Quiltec® I Wipers, Contec®
Quiltec® I Wipers, Contec®
Supplier:  CONTEC, INC. MX
Certificates
undefined
Quiltec® I Wipers, Contec®
Supplier:  CONTEC, INC. MX
Certificates
Specifications
  • Catalog No:
  • 68300-560
  • 89074-012
  • 68310-218
  • 75996-814
  • Peso base:
  • 240 g/m²
  • 240 g/m²
  • 240 g/m²
  • 240 g/m²
  • Absorbancia, extrínseca:
  • 766 ml/m²
  • 766 ml/m²
  • 766 ml/m²
  • 766 ml/m²
  • Absorbancia, intrínseca:
  • 3.19 ml/g
  • 3.19 ml/g
  • 3.19 ml/g
  • 3.19 ml/g
  • Absorbancia, velocidad:
  • <1 s
  • <1 s
  • <1 s
  • <1 s
  • Extraíbles:
  • 0.0028 g/m² using deionized water, 0.0156 g/m² using isopropyl alcohol
  • 0.0028 g/m² using deionized water, 0.0156 g/m² using isopropyl alcohol
  • 0.0028 g/m² using deionized water, 0.0156 g/m² using isopropyl alcohol
  • 0.0028 g/m² using deionized water, 0.0156 g/m² using isopropyl alcohol
  • Particle generation (>=0,5 µm):
  • 6.5×10⁶/m²
  • 6.5×10⁶/m²
  • 6.5×10⁶/m²
  • 6.5×10⁶/m²
  • Iones, cloruro:
  • 1.145 ppm
  • 1.145 ppm
  • 1.145 ppm
  • 1.145 ppm
  • Iones - sodio:
  • 0.696 ppm
  • 0.696 ppm
  • 0.696 ppm
  • 0.696 ppm