Orders LinkContactUsLinkComponent
Buffers, Reference standards, Acidic to Neutral, Ricca Chemical
Buffers, Reference standards, Acidic to Neutral, Ricca Chemical
Buffers, Reference standards, Acidic to Neutral, Ricca Chemical
Buffers, Reference standards, Acidic to Neutral, Ricca Chemical
Buffers, Reference standards, Acidic to Neutral, Ricca Chemical
Colorless pH buffer solutions are certified to ±0.01 pH at 25°C
  • Range of packaging is available
Each 500 ml plastic bottle is labeled with the certified pH value, the NIST SRM number and a pH/temperature conversion.
Certifications: Standardized against solutions whose certification is traceable to the National Institute of Standards and Technology. Meet USP requirements.
Product Family Options
  • pH Accuracy
    pH Buffer Type
    Reference Temperature
    Pack type
    Size
    pH
    Color
  • ±0.01
    HCl
    25 °C
    Poly bottle
    500 ml
    0.50
    Colorless
    Each (500ml)
    Each (500ml) - $753.27
    $753.27
    Each (500ml)
       $753.27
     : 1485-16
  • ±0.01
    HCl/KCl
    25 °C
    Poly bottle
    500 ml
    1.68
    Colorless
    Each (500ml)
    Each (500ml) - $966.55
    $966.55
    Each (500ml)
       $966.55
     : R1492000-500A
  • ±0.01
    HCl/KCl
    25 °C
    Poly bottle
    4 L
    1.68
    Colorless
     : R1492000-4A
  • ±0.01
    HCl/KCl
    25 °C
    Poly bottle
    4 L
    2.00
    Colorless
     : R1493000-4A
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 