MicroSeal® SuperSorb® Sealed-Edge Cleanroom Wipers, Berkshire
Supplier: BERKSHIRE CORP
Certificates
About this item
Bonded 2-ply, 100% knitted polyester wipers feature superior absorbency with a high level of cleanliness and durability.
- Critically low particles, fibers, ions, and extractables
- High abrasion resistance
- Chemically compatible with IPA, Acetone, and other solvents
- Pinsonic design adds surface texture to aid in stubborn residue removal
They are ideal for applications where high concentrations of liquid are used and for cleaning and wiping rough surfaces. Recommended for ISO Class 3 (FED-STD-209E Class 1/M1.5) cleanroom environments, including semiconductor and biomedical cleanroom applications. Steam autoclavable for aseptic environments.
Ordering information: Wipers are laundered and packed in an ISO Class 4 (FED-STD-209E Class 10/M2.5) cleanroom. Wipers come stacked in packs of 100.
Specifications
- Catalog No:
- 89095-084
- 94000-400
- Peso base:
- 253 g/m²
- Absorbancia, extrínseca:
- 967 ml/m²
- Absorbancia, intrínseca:
- 3.8 ml/g
- Absorbancia, velocidad:
- 1 sec.
- Extraíbles:
- 0.022% using deionized water, 0.030% using isopropyl alcohol
- Fibras (>100 µm):
- 0.14/cm²
- Particle generation (>=0,5 µm):
- 0.92 x 10³/m²
- Iones - calcio:
- 0.018 ppm
- Iones, cloruro:
- 0.13 ppm
- Iones - magnesio:
- 0.0093 ppm
- Iones - potasio:
- 0.0063 ppm
- Iones - sodio:
- 0.12 ppm