Orders LinkContactUsLinkComponent
VWR® Total-Range Digital Thermometer
VWR® Total-Range Digital Thermometer
Total-Range Digital Thermometer, –200...1370 °C (–328...2498 °F) when used with appropriate probe
Catalog # 23609-232
Supplier:   VWR International
VWR® Total-Range Digital Thermometer
Catalog # 23609-232
Supplier:  VWR International
VWR® Total-Range Digital Thermometer
VWR® Total-Range Digital Thermometer
Total-Range Digital Thermometer, –200...1370 °C (–328...2498 °F) when used with appropriate probe
Price
$5,570.89
Log in Para disponibilidad
Each (1UN)
Product Details & Documents
Fast-reading, easy-to-use thermometer switches from °C to °F at the touch of a button.
  • Traceable®
  • Reads °C and °F
  • Large display with easily legible digits
  • Memory of minimum and maximum readings
Thermometer recalls minimum and maximum readings from any time period. Alarm function may be programmed in 1° increments to sound when temperature rises above or falls below set points. Count-up timer alarm may be set from 23 hours, 59 minutes, to 1 minute. Unit is constructed of shock-resistant ABS plastic. Water-resistant PVC protective case stops water, fumes, dirt, and dust. Uses all Type-K thermocouple probes. Batteries power unit for one year of routine use.
All Type-K probes may be used with liquids, air/gas, and semi-solids. Stainless steel probe has a 22.9cm (9") stem with a 3mm (1/8") diameter and a 102cm (401/8") cable. Beaded probe is 1.5mm (1/16") in diameter and features a 1.2m (4') fluoropolymer resin cable. The low-temperature stainless steel probe has a diameter of 4.3mm (3/16") and features a 30.5cm (12") stem with a 12.7cm (5") handle. Low-temperature probe is supplied with a 2.4m (8') expandable cable.
Thermometer recalls minimum and maximum readings from any time period. Alarm function may be programmed in 1° increments to sound when temperature rises above or falls below set points. Count-up timer alarm may be set from 23 hours, 59 minutes, to 1 minute. Unit is constructed of shock-resistant ABS plastic. Water-resistant PVC protective case stops water, fumes, dirt, and dust. Uses all Type-K thermocouple probes. Batteries power unit for one year of routine use.
Backlighting button permits reading in dark areas. Unit features a hold button that captures current reading. Microprocessor updates display twice per second.
Certifications: A serial numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate thermometer traceability to standards provided by the National Institute of Standards and Technology.
Ordering information: Supplied with a PVC protective casing with flip-out stand, a fast-response Type-K beaded probe (61161-372), and three AAA alkaline batteries.
Services: VWRCATALYST™ provides convenient and efficient re-calibration services for your digital thermometers with an industry leading 5-day average turn-around time from receipt of your instrument. All the service will be performed by an ISO17025 certified lab. Recalibration, certification and shipping fees are all included in the catalog number cost. Simply order the stated catalog number based on how many calibration points are desired. You will receive a protective box to ship your instrument to Avantor's certified calibration laboratory along with a work order form and a prepaid return shipping label. Upon receipt at the laboratory your instrument will be inspected for any defects and the performance of your instrument will be compared to certified standards. After re-calibration, your calibration report and instrument will be carefully repacked then returned with standard ground shipping. We also provide expedited service options available upon request.
Especificaciones
  • Description:
    Total-Range Digital Thermometer
  • Range:
    –200...1370 °C (–328...2498 °F) when used with appropriate probe
  • Cat. No.:
    23609-232
  • Probe cable length:
    1.2 m (4')
  • W×D×H:
    8.3×3.8×17.8 cm (31/4×11/2 ×7")
  • Temperature resolution:
    0.1° from –199.9° to 1369.9°C (–327.8 to 2497.8°F), 1° rest of range
  • Temperature accuracy:
    ±1°C from –50 to 740°C, ±2°C at ends of range
  • Weight:
    0.2 kg (0.5 lbs.)
Accessories
Información del producto
  • Catalog #
    Ø
    Length
    Availability
    Price
  • Beaded, fast response type-K probe
    Beaded, fast response type-K probe
    61161-372
    1.6 mm (¹/₁₆)"
    Log in Para disponibilidad
    Each (1UN) - $1,115.56