Orders LinkContactUsLinkComponent
PYREX® Watch Glasses, Corning
 :   Corning
PYREX® Watch Glasses, Corning
 :  Corning
PYREX® Watch Glasses, Corning
PYREX® Watch Glasses, Corning
PYREX® borosilicate glass.
  • High chemical durability
  • Prevents contamination of solutions
These watch glasses are molded to the same radius of curvature for convenient stacking. The heavy wall and uniform fire-polished edge provides the mechanical strength. The outside diameter cannot be controlled within close limits and a slight degree of out-of-round can be expected.
They are also used as covers for heated beakers.
 : One package has 12 watch glasses.
Product Family Options
  • Description
    Ø
  • Watch glass
    65 mm (29/16")
    Paquete de 12
    Paquete de 12 - $1,330.00
    $1,330.00
    Case de 12 (12UN)
    Case de 12 (12UN) - $10,483.20
    $10,483.20
    Paquete de 12
       $1,330.00
    Case de 12 (12UN)
       $10,483.20
     : 9985-65
  • Watch glass
    75 mm (3")
    Paquete de 12
    Paquete de 12 - $1,420.00
    $1,420.00
    Case de 12 (12UN)
    Case de 12 (12UN) - $10,689.68
    $10,689.68
    Paquete de 12
       $1,420.00
    Case de 12 (12UN)
       $10,689.68
     : 9985-75
  • Watch glass
    100 mm (315/16")
    Paquete de 12
    Paquete de 12 - $1,700.00
    $1,700.00
    Case de 12 (12UN)
    Case de 12 (12UN) - $1,897.59
    $1,897.59
    Paquete de 12
       $1,700.00
    Case de 12 (12UN)
       $1,897.59
     : 9985-100
  • Watch glass
    150 mm (529/32")
    Paquete de 12
    Paquete de 12 - $3,280.00
    $3,280.00
    Case de 6 (12UN)
    Case de 6 (12UN) - $19,192.78
    $19,192.78
    Paquete de 12
       $3,280.00
    Case de 6 (12UN)
       $19,192.78
     : 9985-150