Accessories for Corning® CellSTACK® Culture Chambers, Sterile, Corning
Catalog # 33500-950
Supplier: CORNING CA
Product available on GSA Advantage®
Specifications
- Descripción:Filling Cap, 3.2 mm (1/8") I.D. Tubing, Female Luer Lock with Male Luer Plug
- Matriz:Polyethylene
- Referencia:33500-950
- Esterilidad:Sterile
- Tamaño de la tapa:33 mm (15/16")
- Nº de proveedor:3282
Specifications
About this item
Caution: This product is not intended to mitigate the presence of microorganisms on surfaces or in the environment, where such organisms can be deleterious to humans or the environment.