ContactUsLinkComponent
KIMAX® Burets with Threaded Closure Fit, Straight Bore PTFE Stopcock, Kimble Chase
KIMAX® Burets with Threaded Closure Fit, Straight Bore PTFE Stopcock, Kimble Chase
Catalog # 89001-648
undefined
KIMAX® Burets with Threaded Closure Fit, Straight Bore PTFE Stopcock, Kimble Chase
Catalog # 89001-648
Supplier Number:  17121H-25
Specifications
  • Tamaño de la tapa:
    13 mm
  • Intervalo de graduación:
    0.1 ml
  • Capacidad:
    25 ml
  • Longitud:
    420 mm
  • Referencia:
    89001-648
  • Nº de proveedor:
    17121H-25
Specifications
About this item
Burets are composed of four replaceable components and incorporate a threaded closure that eliminates accidental dislodging of the stopcock assembly during use
Note: Assembly should be wetted before installation.
The graduated tube has a reinforced top with a lower end accurately tooled to provide precise I.D. fit with a PTFE stopcock assembly. A ground tip fits into the lower end of the stopcock assembly.
Ordering information: Burets include a KIM-KAP® dust cap to secure to the threaded buret body with a positive closure.