PYREX® Micro Burets, Class A, Funnel Top, Precision Bore, Straight [ST] Stopcock, Corning
89091-478
: Corning
Product available on GSA Advantage®
- Ext.Ø:7 mm
- Intervalo de graduación:0.01 ml
- Capacidad:5 ml
- Longitud:735 mm
- Referencia:89091-478
- Tolerancia:±0.01 ml
- Nº de proveedor:2128-5
Micro burets, Class A, PYREX® borosilicate glass.
- Precision bore tubing
- Funnel top
- Tapered tips for smooth flow
- Stopcocks turn freely yet give positive control
Designed for precise analytical methods in microchemistry.
: The capacity tolerance on these burets is established by ASTM E1189 and are calibrated in accordance with ASTM E542.