PYREXPLUS® Safety-Coated Reagent Bottles, Narrow Mouth, Corning
: Corning
PYREX® borosilicate glass, coated.
- Conventional bottle shape
- Autoclavable at 121 °C and resistant to thermal shock
- Used for storage of solutions, ideal for media preparation
Designed with protective PVC coating for longer product life and safety. Protective coating helps prevent glass from shattering and reduces spills. Neck is tooled to increase mechanical strength and for uniform stopper fit.
Not recommended for dry heat sterilization as plastic coating will melt. Can be written on with a standard lab marker, permanent or water-soluble.
: Do not place over direct heat or flames.
Frequently Bought Together